The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017520.1 | NP_001017520.1 | 2 - 3 | 551 | 67 |
NM_004088.3 | NP_004079.3 | 2 - 3 | 551 | 67 | |
GenBank mRNA | AB046378.1 | - | 2 - 3 | 483 | 67 |
AK223317.1 | - | 2 - 3 | 395 | 67 | |
AK313615.1 | - | 2 - 3 | 460 | 67 | |
BC012920.1 | - | 2 - 3 | 424 | 67 |