Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
Chromosome Location
Chr.5: 128257908 - 128538358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001999.3 NP_001990.2 18 - 19 2866 88
XM_017009228.1 XP_016864717.1 17 - 18 3029 88
GenBank mRNA AK300440.1 - 17 - 18 2546 88
BX343658.2 - 6 - 7 714 88
U03272.1 - 18 - 19 2424 88
X62009.1 - 2 - 3 171 88

View Full Product Details