Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
Chromosome Location
Chr.5: 128257908 - 128538358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001999.3 NP_001990.2 19 - 20 2989 68
XM_017009228.1 XP_016864717.1 18 - 19 3152 68
GenBank mRNA AK300440.1 - 18 - 19 2669 68
BX343658.2 - 7 - 8 69
U03272.1 - 19 - 20 2547 68
X62009.1 - 3 - 4 294 68

View Full Product Details