The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001999.3 | NP_001990.2 | 21 - 22 | 3252 | 98 |
XM_017009228.1 | XP_016864717.1 | 20 - 21 | 3415 | 98 | |
GenBank mRNA | AK300440.1 | - | 20 - 21 | 2932 | 98 |
U03272.1 | - | 21 - 22 | 2810 | 98 | |
X62009.1 | - | 5 - 6 | 557 | 98 |