Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
Chromosome Location
Chr.5: 128257908 - 128538358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001999.3 NP_001990.2 23 - 24 3429 121
XM_017009228.1 XP_016864717.1 22 - 23 3592 121
GenBank mRNA AB209735.1 - 2 - 3 182 121
AK300440.1 - 22 - 23 3109 121
U03272.1 - 23 - 24 2987 121
X62009.1 - 7 - 8 734 121

View Full Product Details