Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBN2
Entrez Gene ID
2201
Gene Name
fibrillin 2
Gene Aliases
CCA, DA9, EOMD
Chromosome Location
Chr.5: 128257908 - 128538358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001999.3 NP_001990.2 25 - 26 3784 76
XM_017009228.1 XP_016864717.1 24 - 25 3947 76
GenBank mRNA AB209735.1 - 4 - 5 537 76
AK300440.1 - 24 - 25 3464 76
BG009811.1 - 1 - 2 153 76
CN300894.1 - 1 - 2 65 76
U03272.1 - 25 - 26 3342 76
X62009.1 - 9 - 10 1089 76

View Full Product Details