The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258450.1 | NP_001245379.1 | 6 - 7 | 1459 | 122 |
NM_001258451.1 | NP_001245380.1 | 6 - 7 | 1530 | 122 | |
NM_004203.4 | NP_004194.3 | 6 - 7 | 1666 | 122 | |
NM_182687.2 | NP_872629.1 | 6 - 7 | 1666 | 122 | |
XM_011522734.2 | XP_011521036.1 | 6 - 7 | 1627 | 122 | |
XM_011522735.2 | XP_011521037.1 | 6 - 7 | 1453 | 122 | |
XM_011522736.2 | XP_011521038.1 | 5 - 6 | 1460 | 122 | |
GenBank mRNA | AK097642.1 | - | 6 - 7 | 1415 | 122 |
AK098452.1 | - | 6 - 7 | 1518 | 122 | |
AK294379.1 | - | 5 - 6 | 1427 | 122 | |
AK301926.1 | - | 6 - 7 | 1335 | 122 | |
AK303005.1 | - | 6 - 7 | 1544 | 122 | |
AK309964.1 | - | 5 - 6 | 1460 | 122 | |
BC121161.1 | - | 5 - 6 | 1354 | 122 | |
BC121162.1 | - | 5 - 6 | 1354 | 122 | |
BC130011.1 | - | 5 - 6 | 1354 | 122 | |
BC153823.1 | - | 5 - 6 | 1354 | 122 | |
U56816.1 | - | 5 - 6 | 1370 | 122 |