The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001140.3 | NP_001131.3 | 11 - 12 | 1555 | 64 |
GenBank mRNA | AK290309.1 | - | 11 - 12 | 1568 | 64 |
AK296792.1 | - | 9 - 10 | 1212 | 64 | |
AK296821.1 | - | 12 - 13 | 1640 | 64 | |
AK296904.1 | - | 11 - 12 | 1451 | 64 | |
AK312557.1 | - | 11 - 12 | 1568 | 64 | |
AK316126.1 | - | 11 - 12 | 1711 | 64 | |
BC029032.1 | - | 11 - 12 | 1555 | 64 | |
M23892.1 | - | 11 - 12 | 1544 | 64 |