The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014228.4 | NP_055043.2 | 1 - 2 | 499 | 92 |
XM_017009767.1 | XP_016865256.1 | 1 - 2 | 295 | 92 | |
XM_017009768.1 | XP_016865257.1 | 1 - 2 | 133 | 92 | |
XM_017009769.1 | XP_016865258.1 | 1 - 2 | 276 | 92 | |
XM_017009770.1 | XP_016865259.1 | 1 - 2 | 73 | 92 | |
GenBank mRNA | AK096607.1 | - | 1 - 2 | 403 | 92 |
AK308895.1 | - | 1 - 2 | 275 | 92 | |
AK312309.1 | - | 1 - 2 | 279 | 92 | |
BC093785.1 | - | 1 - 2 | 108 | 92 | |
BC113425.1 | - | 1 - 2 | 108 | 92 | |
DC321993.1 | - | 1 - 2 | 499 | 92 |