Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM1
Entrez Gene ID
8736
Gene Name
myomesin 1
Gene Aliases
SKELEMIN
Chromosome Location
Chr.18: 3066807 - 3220108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003803.3 NP_003794.3 15 - 16 2547 75
NM_019856.1 NP_062830.1 15 - 16 2547 75
XM_017026062.1 XP_016881551.1 15 - 16 2310 75
GenBank mRNA AJ621424.1 - 15 - 16 2547 75
AK125322.1 - 16 - 17 2547 75
BC116183.1 - 15 - 16 2288 75
X69090.1 - 13 - 14 1919 75

View Full Product Details