Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHX34
Entrez Gene ID
9704
Gene Name
DEAH-box helicase 34
Gene Aliases
DDX34, HRH1
Chromosome Location
Chr.19: 47349281 - 47382704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AA454494.1 - 2 - 3 170 132

View Full Product Details