Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VILL
Entrez Gene ID
50853
Gene Name
villin like
Gene Aliases
-
Chromosome Location
Chr.3: 37987974 - 38007188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015873.3 NP_056957.3 17 - 18 2292 65
XM_005265191.3 XP_005265248.1 18 - 19 2400 65
XM_005265192.4 XP_005265249.1 18 - 19 3535 65
XM_006713184.3 XP_006713247.1 19 - 20 2400 65
XM_011533770.2 XP_011532072.1 18 - 19 2556 65
GenBank mRNA BC000243.1 - 14 - 15 1828 65
BC004300.1 - 18 - 19 2322 65
D88154.1 - 13 - 14 1698 65
KU178619.1 - 13 - 14 1696 65
KU178620.1 - 12 - 13 1550 65

View Full Product Details