The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015873.3 | NP_056957.3 | 11 - 12 | 1410 | 72 |
XM_005265191.3 | XP_005265248.1 | 12 - 13 | 1518 | 72 | |
XM_005265192.4 | XP_005265249.1 | 12 - 13 | 2653 | 72 | |
XM_006713184.3 | XP_006713247.1 | 13 - 14 | 1518 | 72 | |
XM_011533770.2 | XP_011532072.1 | 12 - 13 | 1674 | 72 | |
XM_011533771.1 | XP_011532073.1 | 12 - 13 | 1674 | 72 | |
XM_011533773.2 | XP_011532075.1 | 12 - 13 | 1675 | 72 | |
GenBank mRNA | AK309304.1 | - | 9 - 10 | 1300 | 72 |
BC000243.1 | - | 8 - 9 | 946 | 72 | |
BC004300.1 | - | 12 - 13 | 1440 | 72 | |
KU178619.1 | - | 7 - 8 | 814 | 72 | |
KU178620.1 | - | 7 - 8 | 814 | 72 |