The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161581.1 | NP_001155053.1 | 10 - 11 | 1516 | 54 |
NM_015426.4 | NP_056241.3 | 10 - 11 | 1445 | 54 | |
XM_011533560.1 | XP_011531862.1 | 9 - 10 | 1068 | 54 | |
GenBank mRNA | AK075289.1 | - | 10 - 11 | 1160 | 54 |
AK315688.1 | - | 10 - 11 | 1167 | 54 | |
AL117629.1 | - | 5 - 6 | 484 | 54 | |
BC007417.2 | - | 9 - 10 | 1095 | 54 | |
BC119692.2 | - | 10 - 11 | 1155 | 54 | |
BC119693.2 | - | 10 - 11 | 1231 | 54 |