The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195422.1 | NP_001182351.1 | 7 - 8 | 1063 | 134 |
NM_032620.3 | NP_116009.2 | 7 - 8 | 1070 | 134 | |
NM_133644.3 | NP_598399.2 | 6 - 7 | 1166 | 134 | |
GenBank mRNA | AF360742.1 | - | 7 - 8 | 1032 | 134 |
AK027606.1 | - | 7 - 8 | 1017 | 134 | |
AK291929.1 | - | 7 - 8 | 1014 | 134 | |
AK297953.1 | - | 7 - 8 | 1045 | 134 | |
AK298485.1 | - | - | 727 | 134 | |
AY078987.1 | - | 6 - 7 | 1128 | 134 | |
AY078988.1 | - | 7 - 8 | 1032 | 134 | |
BC017207.2 | - | 7 - 8 | 1002 | 134 | |
BC019261.1 | - | 7 - 8 | 1011 | 134 |