Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PDZK1
- Entrez Gene ID
-
5174
- Gene Name
- PDZ domain containing 1
- Gene Aliases
-
CAP70, CLAMP, NHERF-3, NHERF3, PDZD1
- Chromosome Location
-
Chr.1: 145670852 - 145707507 on Build GRCh38
- Gene Symbol
- PDZK1P1
- Entrez Gene ID
-
100034743
- Gene Name
- PDZ domain containing 1 pseudogene 1
- Gene Aliases
-
OTTHUMT00000038524, PDZK1P2
- Chromosome Location
-
Chr.1: 147993862 - 148014956 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111936.1 |
|
6 - 7 |
989 |
99 |
GenBank mRNA |
AK126719.1 |
- |
5 - 6 |
2358 |
99 |
View Full Product Details