Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFHX3
Entrez Gene ID
463
Gene Name
zinc finger homeobox 3
Gene Aliases
ATBF1, ATBT, ZNF927
Chromosome Location
Chr.16: 72782885 - 73058635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164766.1 NP_001158238.1 3 - 4 832 134
NM_006885.3 NP_008816.3 4 - 5 4118 134
XM_017023251.1 XP_016878740.1 3 - 4 9828 134
GenBank mRNA BC029653.1 - 3 - 4 1095 134
D10250.1 - 3 - 4 832 134
L32832.1 - 4 - 5 4118 134

View Full Product Details