The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019601.3 | NP_062547.1 | 7 - 8 | 1113 | 102 |
GenBank mRNA | AK026431.1 | - | 7 - 8 | 1131 | 102 |
AK126105.1 | - | 5 - 6 | 1962 | 102 | |
BC033107.1 | - | 7 - 8 | 1113 | 102 |