The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318245.1 | NP_001305174.1 | 24 - 25 | 4485 | 121 |
NM_032608.6 | NP_115997.5 | 24 - 25 | 4482 | 121 | |
XM_011530458.2 | XP_011528760.1 | 25 - 26 | 4621 | 121 | |
XM_011530459.2 | XP_011528761.1 | 25 - 26 | 4592 | 121 | |
XM_011530460.2 | XP_011528762.1 | 24 - 25 | 4465 | 121 | |
XM_011530461.2 | XP_011528763.1 | 25 - 26 | 4621 | 121 | |
XM_011530464.2 | XP_011528766.1 | 25 - 26 | 4618 | 121 | |
XM_011530465.2 | XP_011528767.1 | 24 - 25 | 4378 | 121 | |
XM_011530466.2 | XP_011528768.1 | 25 - 26 | 4622 | 121 | |
XM_017029012.1 | XP_016884501.1 | 25 - 26 | 4621 | 121 | |
XM_017029013.1 | XP_016884502.1 | 25 - 26 | 4669 | 121 | |
XM_017029014.1 | XP_016884503.1 | 25 - 26 | 4621 | 121 | |
XM_017029015.1 | XP_016884504.1 | 24 - 25 | 4492 | 121 | |
XM_017029016.1 | XP_016884505.1 | 13 - 14 | 1745 | 121 | |
XM_017029017.1 | XP_016884506.1 | 23 - 24 | 4346 | 121 | |
GenBank mRNA | AB042648.2 | - | 20 - 21 | 2794 | 121 |
AB075376.1 | - | 24 - 25 | 4482 | 121 | |
AJ310931.2 | - | 24 - 25 | 4473 | 121 | |
AY077700.1 | - | 24 - 25 | 4395 | 121 | |
BC144596.1 | - | 22 - 23 | 4137 | 121 | |
BC144597.1 | - | 23 - 24 | 4282 | 121 | |
BC150626.1 | - | 23 - 24 | 4282 | 121 |