Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr.22: 25742133 - 26063847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318245.1 NP_001305174.1 35 - 36 5897 63
NM_032608.6 NP_115997.5 35 - 36 5894 63
XM_011530458.2 XP_011528760.1 36 - 37 6033 63
XM_011530459.2 XP_011528761.1 36 - 37 6004 63
XM_011530460.2 XP_011528762.1 35 - 36 5877 63
XM_011530461.2 XP_011528763.1 36 - 37 6033 63
XM_011530464.2 XP_011528766.1 36 - 37 6030 63
XM_011530465.2 XP_011528767.1 35 - 36 5790 63
XM_011530466.2 XP_011528768.1 36 - 37 6034 63
XM_017029012.1 XP_016884501.1 36 - 37 6033 63
XM_017029013.1 XP_016884502.1 36 - 37 6081 63
XM_017029014.1 XP_016884503.1 36 - 37 6030 63
XM_017029015.1 XP_016884504.1 35 - 36 5901 63
XM_017029016.1 XP_016884505.1 24 - 25 3157 63
XM_017029017.1 XP_016884506.1 34 - 35 5758 63
GenBank mRNA AB042648.2 - 31 - 32 4206 63
AB075376.1 - 35 - 36 5894 63
AJ310931.2 - 35 - 36 5885 63
AY077700.1 - 35 - 36 5807 63
BC144596.1 - 33 - 34 5549 63
BC144597.1 - 34 - 35 5691 63
BC150626.1 - 34 - 35 5691 63

View Full Product Details