The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000111.2 | NP_000102.1 | 14 - 15 | 1800 | 161 |
XM_011515867.2 | XP_011514169.1 | 14 - 15 | 1781 | 161 | |
GenBank mRNA | AK300651.1 | - | 8 - 9 | 828 | 161 |
BC025671.1 | - | 14 - 15 | 1774 | 161 | |
BX640837.1 | - | 14 - 15 | 1743 | 161 | |
L02785.1 | - | 14 - 15 | 1773 | 161 |