The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256510.1 | NP_001243439.1 | 5 - 6 | 748 | 111 |
NM_001256511.1 | NP_001243440.1 | 5 - 6 | 629 | 111 | |
NM_001256512.1 | NP_001243441.1 | 5 - 6 | 611 | 111 | |
NM_001256513.1 | NP_001243442.1 | 5 - 6 | 539 | 111 | |
NM_003143.2 | NP_003134.1 | 5 - 6 | 446 | 111 | |
NR_046269.1 | 6 - 7 | 496 | 111 | ||
XM_005250048.4 | XP_005250105.1 | 5 - 6 | 458 | 111 | |
XM_005250049.4 | XP_005250106.1 | 5 - 6 | 497 | 111 | |
XM_005250050.4 | XP_005250107.1 | 5 - 6 | 548 | 111 | |
XM_005250051.4 | XP_005250108.1 | 5 - 6 | 574 | 111 | |
GenBank mRNA | AF277319.1 | - | 5 - 6 | 392 | 111 |
AK294393.1 | - | 5 - 6 | 402 | 111 | |
AK313033.1 | - | 5 - 6 | 459 | 111 | |
BC000895.1 | - | 5 - 6 | 748 | 111 | |
BF131408.1 | - | 6 - 7 | 426 | 112 | |
BP308841.1 | - | 4 - 5 | 315 | 111 | |
BQ959702.1 | - | 5 - 6 | 511 | 111 | |
M94556.1 | - | 5 - 6 | 391 | 111 |