The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017006967.1 | XP_016862456.1 | 1 - 2 | 153 | 138 |
GenBank mRNA | AK292063.1 | - | 1 - 2 | 111 | 138 |
AL706210.1 | - | 1 - 2 | 159 | 138 | |
BC018141.1 | - | 1 - 2 | 98 | 138 | |
CD014089.1 | - | 1 - 2 | 148 | 138 | |
DA426203.1 | - | 1 - 2 | 151 | 138 | |
M16541.1 | - | 1 - 2 | 120 | 138 |