Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGLL
Entrez Gene ID
11343
Gene Name
monoglyceride lipase
Gene Aliases
HU-K5, HUK5, MAGL, MGL
Chromosome Location
Chr.3: 127689062 - 127823250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003794.2 NP_001003794.1 4 - 5 559 64
NM_001256585.1 NP_001243514.1 4 - 5 966 64
NM_007283.6 NP_009214.1 4 - 5 966 64
XM_011512377.1 XP_011510679.1 4 - 5 938 64
XM_011512378.1 XP_011510680.1 4 - 5 559 64
XM_011512379.2 XP_011510681.1 5 - 6 675 64
XM_011512382.1 XP_011510684.1 3 - 4 501 64
XM_011512383.2 XP_011510685.1 5 - 6 630 64
XM_017005662.1 XP_016861151.1 4 - 5 559 64
XM_017005663.1 XP_016861152.1 5 - 6 630 64
XM_017005664.1 XP_016861153.1 5 - 6 1127 64
XM_017005665.1 XP_016861154.1 3 - 4 689 64
XM_017005666.1 XP_016861155.1 4 - 5 559 64
GenBank mRNA AJ270950.1 - 3 - 4 372 64
AK091314.1 - 4 - 5 1269 64
AK304844.1 - 4 - 5 435 64
AK310874.1 - 4 - 5 494 64
AK315529.1 - 4 - 5 606 64
AY172950.1 - 2 - 3 792 64
BC000551.2 - 4 - 5 551 64
BC006230.2 - 4 - 5 921 64
BE302457.1 - 4 - 5 508 64
BG035493.1 - 1 - 2 142 64
BP307995.1 - 4 - 5 537 64
BX403703.2 - 4 - 5 478 64
CR456835.1 - 3 - 4 402 64
EU660904.1 - 3 - 4 372 64
U67963.1 - 4 - 5 552 64

View Full Product Details