The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014937.3 | NP_055752.1 | 5 - 6 | 828 | 145 |
XM_006717720.3 | XP_006717783.1 | 5 - 6 | 3215 | 145 | |
XM_011539524.2 | XP_011537826.1 | 5 - 6 | 591 | 145 | |
XM_011539525.2 | XP_011537827.1 | 6 - 7 | 3298 | 145 | |
XM_011539527.2 | XP_011537829.2 | 4 - 5 | 593 | 145 | |
XM_011539528.2 | XP_011537830.1 | 4 - 5 | 715 | 145 | |
XM_017015935.1 | XP_016871424.1 | 5 - 6 | 582 | 145 | |
XM_017015936.1 | XP_016871425.1 | 8 - 9 | 1216 | 145 | |
GenBank mRNA | AB023183.1 | - | 5 - 6 | 778 | 145 |
AK307758.1 | - | 5 - 6 | 818 | 145 | |
BC067820.1 | - | 5 - 6 | 789 | 145 | |
BC111493.1 | - | 5 - 6 | 739 | 145 |