The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014937.3 | NP_055752.1 | 8 - 9 | 1259 | 79 |
XM_006717720.3 | XP_006717783.1 | 8 - 9 | 3646 | 79 | |
XM_011539524.2 | XP_011537826.1 | 8 - 9 | 1022 | 79 | |
XM_011539525.2 | XP_011537827.1 | 9 - 10 | 3729 | 79 | |
XM_011539527.2 | XP_011537829.2 | 7 - 8 | 1024 | 79 | |
XM_011539528.2 | XP_011537830.1 | 7 - 8 | 1146 | 79 | |
XM_017015935.1 | XP_016871424.1 | 8 - 9 | 1013 | 79 | |
XM_017015936.1 | XP_016871425.1 | 11 - 12 | 1647 | 79 | |
GenBank mRNA | AB023183.1 | - | 8 - 9 | 1209 | 79 |
AK307758.1 | - | 8 - 9 | 1250 | 79 | |
AL137528.1 | - | 2 - 3 | 224 | 79 | |
BC067820.1 | - | 8 - 9 | 1220 | 79 | |
BC111493.1 | - | 8 - 9 | 1170 | 79 |