Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
Chromosome Location
Chr.1: 186375758 - 186421375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164245.1 NP_001157717.1 1 - 2 217 85
NM_001164246.1 NP_001157718.1 1 - 2 217 85
NM_017847.5 NP_060317.3 1 - 2 217 85
XM_011509660.1 XP_011507962.1 1 - 2 216 85
XM_011509661.1 XP_011507963.1 1 - 2 216 85
XM_017001543.1 XP_016857032.1 1 - 2 216 85
GenBank mRNA AK000512.1 - 1 - 2 137 85
AK298107.1 - 1 - 2 153 85
AY854248.1 - 1 - 2 110 85
BC003397.1 - 1 - 2 50 85
DB116803.1 - 1 - 2 218 85

View Full Product Details