Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
Chromosome Location
Chr.1: 186375758 - 186421375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164245.1 NP_001157717.1 7 - 8 847 72
NM_001164246.1 NP_001157718.1 6 - 7 751 72
NM_017847.5 NP_060317.3 7 - 8 847 72
XM_011509660.1 XP_011507962.1 7 - 8 846 72
XM_011509661.1 XP_011507963.1 7 - 8 846 72
XM_017001543.1 XP_016857032.1 7 - 8 846 72
GenBank mRNA AK000512.1 - 7 - 8 767 72
AK026086.1 - 5 - 6 417 72
AK298107.1 - 6 - 7 687 72
AY854248.1 - 7 - 8 740 72
BC003397.1 - 7 - 8 680 72
DQ323046.1 - 6 - 7 611 72
DQ529299.1 - 6 - 7 611 72

View Full Product Details