The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001154.3 | NP_001145.1 | 7 - 8 | 639 | 63 |
GenBank mRNA | AK312644.1 | - | 7 - 8 | 639 | 63 |
AY577779.1 | - | 5 - 6 | 475 | 63 | |
BC001429.2 | - | 6 - 7 | 544 | 63 | |
BC004993.1 | - | 7 - 8 | 597 | 63 | |
BC012804.2 | - | 7 - 8 | 591 | 63 | |
BC012822.2 | - | 7 - 8 | 591 | 63 | |
BC018671.1 | - | 7 - 8 | 631 | 63 | |
BC032093.1 | - | 7 - 8 | 628 | 63 | |
CR536522.1 | - | 5 - 6 | 474 | 63 | |
D00172.1 | - | 7 - 8 | 609 | 63 | |
EU794590.1 | - | 7 - 8 | 639 | 63 | |
J03745.1 | - | 7 - 8 | 633 | 63 | |
M18366.1 | - | 6 - 7 | 486 | 63 | |
M19384.1 | - | 7 - 8 | 624 | 63 | |
M21731.1 | - | 7 - 8 | 616 | 63 | |
X12454.1 | - | 7 - 8 | 602 | 63 |