The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080424.1 | NP_001073893.1 | 17 - 18 | 4536 | 76 |
XM_005256549.3 | XP_005256606.1 | 18 - 19 | 4688 | 76 | |
XM_005256550.4 | XP_005256607.1 | 17 - 18 | 4530 | 76 | |
XM_005256551.3 | XP_005256608.1 | 17 - 18 | 4569 | 76 | |
XM_005256552.4 | XP_005256609.1 | 17 - 18 | 5542 | 76 | |
XM_006721483.3 | XP_006721546.1 | 18 - 19 | 4633 | 76 | |
XM_011523750.2 | XP_011522052.1 | 18 - 19 | 5893 | 76 | |
XM_011523752.2 | XP_011522054.1 | 17 - 18 | 4533 | 76 | |
XM_017024381.1 | XP_016879870.1 | 17 - 18 | 5979 | 76 | |
GenBank mRNA | AB002344.2 | - | 17 - 18 | 4541 | 76 |
BC009994.1 | - | 7 - 8 | 1165 | 76 |