Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENDOV
Entrez Gene ID
284131
Gene Name
endonuclease V
Gene Aliases
-
Chromosome Location
Chr.17: 80415117 - 80438086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164637.2 NP_001158109.1 6 - 7 607 94
NM_001164638.1 NP_001158110.1 6 - 7 607 94
NM_173627.4 NP_775898.2 7 - 8 742 94
XM_005257244.3 XP_005257301.1 7 - 8 792 94
XM_005257246.3 XP_005257303.1 6 - 7 609 94
XM_006721837.3 XP_006721900.1 7 - 8 792 94
XM_011524662.1 XP_011522964.1 7 - 8 825 94
XM_011524663.1 XP_011522965.1 7 - 8 825 94
XM_011524669.2 XP_011522971.1 7 - 8 825 94
XM_017024512.1 XP_016880001.1 7 - 8 920 94
XM_017024514.1 XP_016880003.1 6 - 7 590 94
XM_017024515.1 XP_016880004.1 5 - 6 550 94
XM_017024516.1 XP_016880005.1 4 - 5 453 94
GenBank mRNA AK092539.1 - 6 - 7 607 94
AK096344.1 - 7 - 8 742 94
AK096802.1 - 5 - 6 475 94
BC064545.1 - 6 - 7 590 94
BU553848.1 - 3 - 4 94
BX647411.1 - 4 - 5 470 94

View Full Product Details