The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016510.5 | NP_057594.4 | 1 - 2 | 222 | 62 |
GenBank mRNA | AB209458.2 | - | 1 - 2 | 223 | 62 |
BC000586.2 | - | 1 - 2 | 38 | 62 | |
BC007891.2 | - | 1 - 2 | 107 | 62 | |
DN995828.1 | - | 1 - 2 | 162 | 62 |