The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024574.3 | NP_078850.3 | 1 - 2 | 528 | 126 |
GenBank mRNA | AK289615.1 | - | 1 - 2 | 436 | 126 |
AY358821.1 | - | 1 - 2 | 100 | 126 | |
BC019351.1 | - | 1 - 2 | 424 | 126 |