The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000351.5 | NP_000342.2 | 3 - 4 | 522 | 89 |
NM_001320750.1 | NP_001307679.1 | 4 - 5 | 746 | 89 | |
NM_001320751.1 | NP_001307680.1 | 5 - 6 | 740 | 89 | |
NM_001320752.1 | NP_001307681.1 | 4 - 5 | 586 | 89 | |
NM_001320753.1 | NP_001307682.1 | 3 - 4 | 364 | 89 | |
NM_001320754.1 | NP_001307683.1 | 3 - 4 | 512 | 89 | |
GenBank mRNA | AK314034.1 | - | 3 - 4 | 462 | 89 |
BC075030.2 | - | 3 - 4 | 298 | 89 | |
J04964.1 | - | 3 - 4 | 480 | 89 | |
M16505.1 | - | 3 - 4 | 497 | 89 |