Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB42
Entrez Gene ID
2038
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
PA, SPH5
Chromosome Location
Chr.15: 43197227 - 43225856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000119.2 NP_000110.2 2 - 3 584 62
NM_001114134.1 NP_001107606.1 2 - 3 494 62
XM_005254225.1 XP_005254282.1 2 - 3 331 62
XM_011521349.2 XP_011519651.1 3 - 4 1312 62
XM_011521350.2 XP_011519652.1 4 - 5 1044 62
XM_011521351.2 XP_011519653.1 4 - 5 906 62
XM_011521352.2 XP_011519654.1 2 - 3 523 62
XM_011521353.2 XP_011519655.1 2 - 3 522 62
GenBank mRNA AK297146.1 - 2 - 3 268 62
BC096093.4 - 2 - 3 273 62
BC096094.2 - 2 - 3 273 62
BC099627.1 - 2 - 3 273 62
M29399.1 - 2 - 3 381 62
M30646.1 - 2 - 3 421 62
M30647.1 - 2 - 3 511 62
M60298.1 - 2 - 3 743 62

View Full Product Details