The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003054.4 | NP_003045.2 | 3 - 4 | 623 | 67 |
GenBank mRNA | AK314997.1 | - | 3 - 4 | 576 | 67 |
BC030593.1 | - | 3 - 4 | 608 | 67 | |
BC108927.1 | - | 2 - 3 | 488 | 67 | |
BC108928.2 | - | 2 - 3 | 488 | 67 | |
L09118.1 | - | 3 - 4 | 565 | 67 | |
L14269.1 | - | 3 - 4 | 493 | 67 | |
L23205.1 | - | 3 - 4 | 573 | 67 | |
X71354.1 | - | 3 - 4 | 523 | 67 |