The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003054.4 | NP_003045.2 | 6 - 7 | 865 | 89 |
GenBank mRNA | AK314997.1 | - | 6 - 7 | 818 | 89 |
BC030593.1 | - | 5 - 6 | 1123 | 89 | |
BC108927.1 | - | 5 - 6 | 730 | 89 | |
BC108928.2 | - | 5 - 6 | 730 | 89 | |
L09118.1 | - | 6 - 7 | 807 | 89 | |
L14269.1 | - | 6 - 7 | 735 | 89 | |
L23205.1 | - | 6 - 7 | 815 | 89 | |
X71354.1 | - | 6 - 7 | 765 | 89 |