The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004704.4 | NP_004695.1 | 10 - 11 | 1045 | 66 |
GenBank mRNA | AJ001340.1 | - | 10 - 11 | 1005 | 66 |
AK313696.1 | - | 10 - 11 | 1045 | 66 | |
BC001113.1 | - | 10 - 11 | 1028 | 66 | |
BC009879.2 | - | 10 - 11 | 1010 | 66 | |
BC010048.1 | - | 10 - 11 | 1028 | 66 | |
BC021032.1 | - | 10 - 11 | 1028 | 66 | |
BC023662.2 | - | 10 - 11 | 976 | 66 |