Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRP9
Entrez Gene ID
9136
Gene Name
ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
RNU3IP2, U3-55K
Chromosome Location
Chr.3: 51933426 - 51941941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004704.4 NP_004695.1 11 - 12 1109 73
GenBank mRNA AJ001340.1 - 11 - 12 1069 73
AK313696.1 - 11 - 12 1109 73
BC001113.1 - 11 - 12 1092 73
BC009879.2 - 11 - 12 1074 73
BC010048.1 - 11 - 12 1092 73
BC021032.1 - 11 - 12 1092 73
BC023662.2 - 11 - 12 1040 73

View Full Product Details