The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005862.2 | NP_005853.2 | 28 - 29 | 3361 | 117 |
XM_011512329.2 | XP_011510631.1 | 30 - 31 | 3565 | 117 | |
XM_011512331.2 | XP_011510633.1 | 29 - 30 | 3495 | 117 | |
XM_017005523.1 | XP_016861012.1 | 27 - 28 | 3239 | 117 | |
XM_017005524.1 | XP_016861013.1 | 28 - 29 | 3435 | 117 | |
XM_017005525.1 | XP_016861014.1 | 16 - 17 | 1933 | 117 | |
GenBank mRNA | AB210023.1 | - | 28 - 29 | 3326 | 117 |
AK304381.1 | - | 17 - 18 | 1901 | 117 | |
BC017735.1 | - | 5 - 6 | 711 | 117 | |
BC064699.1 | - | 28 - 29 | 3336 | 117 | |
CR749247.1 | - | 30 - 31 | 3633 | 117 | |
Z75330.1 | - | 28 - 29 | 3469 | 117 |