Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A3
Entrez Gene ID
6531
Gene Name
solute carrier family 6 member 3
Gene Aliases
DAT, DAT1, PKDYS
Chromosome Location
Chr.5: 1392790 - 1445428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044.4 NP_001035.1 8 - 9 1287 80
GenBank mRNA BC132977.1 - 7 - 8 1212 80
BC133003.1 - 7 - 8 1212 80
EF174603.1 - 8 - 9 1289 80
L24178.1 - 7 - 8 1180 80
M95167.1 - 8 - 9 1289 80
S44626.1 - 7 - 8 1186 80
S46955.1 - 8 - 9 1262 80

View Full Product Details