The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000377.2 | NP_000368.1 | 7 - 8 | 792 | 103 |
XM_011543977.2 | XP_011542279.1 | 7 - 8 | 797 | 103 | |
XM_017029786.1 | XP_016885275.1 | 7 - 8 | 836 | 103 | |
GenBank mRNA | BC002961.1 | - | 7 - 8 | 771 | 103 |
BC012738.2 | - | 7 - 8 | 763 | 103 | |
U12707.1 | - | 7 - 8 | 769 | 103 | |
U19927.1 | - | 7 - 8 | 759 | 103 |