The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000489.4 | NP_000480.3 | 26 - 27 | 6376 | 154 |
NM_138270.3 | NP_612114.2 | 25 - 26 | 6262 | 154 | |
XM_005262153.4 | XP_005262210.2 | 26 - 27 | 6373 | 154 | |
XM_005262154.4 | XP_005262211.2 | 27 - 28 | 6289 | 154 | |
XM_005262155.4 | XP_005262212.2 | 26 - 27 | 6477 | 154 | |
XM_005262156.4 | XP_005262213.2 | 25 - 26 | 6229 | 154 | |
XM_005262157.4 | XP_005262214.2 | 26 - 27 | 6172 | 154 | |
XM_006724666.3 | XP_006724729.1 | 25 - 26 | 6259 | 154 | |
XM_006724667.3 | XP_006724730.1 | 24 - 25 | 6115 | 154 | |
XM_017029601.1 | XP_016885090.1 | 27 - 28 | 6286 | 154 | |
XM_017029602.1 | XP_016885091.1 | 26 - 27 | 6457 | 154 | |
XM_017029603.1 | XP_016885092.1 | 25 - 26 | 6226 | 154 | |
XM_017029604.1 | XP_016885093.1 | 26 - 27 | 6175 | 154 | |
XM_017029605.1 | XP_016885094.1 | 27 - 28 | 6390 | 154 | |
XM_017029606.1 | XP_016885095.1 | 25 - 26 | 6363 | 154 | |
XM_017029607.1 | XP_016885096.1 | 25 - 26 | 6360 | 154 | |
XM_017029608.1 | XP_016885097.1 | 24 - 25 | 6112 | 154 | |
XM_017029609.1 | XP_016885098.1 | 26 - 27 | 6276 | 154 | |
XM_017029610.1 | XP_016885099.1 | 26 - 27 | 6273 | 154 | |
XM_017029611.1 | XP_016885100.1 | 25 - 26 | 6028 | 154 | |
GenBank mRNA | AB102641.1 | - | 26 - 27 | 6108 | 154 |
AK295107.1 | - | 9 - 10 | 1228 | 154 | |
AK296959.1 | - | 5 - 6 | 611 | 154 | |
BX647222.1 | - | 1 - 2 | 3180 | 154 | |
U09820.1 | - | 18 - 19 | 3587 | 154 | |
U72936.2 | - | 27 - 28 | 6445 | 154 | |
U72937.2 | - | 26 - 27 | 6323 | 154 | |
U72938.2 | - | 25 - 26 | 6209 | 154 |