The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014467.2 | NP_055282.1 | 5 - 6 | 1014 | 96 |
GenBank mRNA | AF060567.1 | - | 5 - 6 | 952 | 96 |
AF393649.1 | - | 5 - 6 | 873 | 96 | |
AK075462.1 | - | 5 - 6 | 962 | 96 | |
BC020733.1 | - | 5 - 6 | 789 | 96 |