Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VCP
Entrez Gene ID
7415
Gene Name
valosin containing protein
Gene Aliases
ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97
Chromosome Location
Chr.9: 35056068 - 35072742 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007126.3 NP_009057.1 12 - 13 1865 82
GenBank mRNA AF100752.1 - 12 - 13 1688 82
AK307735.1 - 12 - 13 1680 82
AK312310.1 - 12 - 13 1747 82
AL137377.1 - 3 - 4 351 82
BC007562.1 - 8 - 9 991 82
BC110913.1 - 12 - 13 1675 82
BC121794.1 - 12 - 13 1672 82
BC122550.1 - 11 - 12 1750 82
FJ224344.1 - 12 - 13 1688 82
FJ224352.1 - 12 - 13 1688 82

View Full Product Details