Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
Chromosome Location
Chr.11: 63998554 - 64166652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014067.3 NP_054786.2 3 - 4 636 80
XM_005273939.4 XP_005273996.1 3 - 4 1175 80
GenBank mRNA AF086206.1 - 3 - 4 256 80
AF202922.2 - 3 - 4 569 80
BC000270.1 - 3 - 4 584 80
BC003188.1 - 3 - 4 390 80
BC007297.1 - 3 - 4 584 80
BC008316.1 - 3 - 4 584 80

View Full Product Details