The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039469.2 | NP_001034558.2 | 11 - 12 | 1677 | 149 |
NM_001163296.1 | NP_001156768.1 | 11 - 12 | 1677 | 149 | |
NM_001163297.1 | NP_001156769.1 | 11 - 12 | 1677 | 149 | |
NM_004954.4 | NP_004945.4 | 11 - 12 | 1677 | 149 | |
NM_017490.3 | NP_059672.2 | 11 - 12 | 1494 | 149 | |
XM_011544789.1 | XP_011543091.1 | 12 - 13 | 1568 | 149 | |
XM_011544790.1 | XP_011543092.1 | 12 - 13 | 1568 | 149 | |
XM_011544791.1 | XP_011543093.1 | 12 - 13 | 1568 | 149 | |
XM_011544792.1 | XP_011543094.1 | 12 - 13 | 1568 | 149 | |
XM_011544793.1 | XP_011543095.1 | 12 - 13 | 1568 | 149 | |
XM_011544794.1 | XP_011543096.1 | 12 - 13 | 1568 | 149 | |
XM_011544795.1 | XP_011543097.1 | 12 - 13 | 1568 | 149 | |
XM_011544796.1 | XP_011543098.1 | 12 - 13 | 1568 | 149 | |
XM_011544797.1 | XP_011543099.1 | 12 - 13 | 1568 | 149 | |
XM_011544798.1 | XP_011543100.1 | 12 - 13 | 1568 | 149 | |
XM_017017310.1 | XP_016872799.1 | 11 - 12 | 1650 | 149 | |
XM_017017311.1 | XP_016872800.1 | 12 - 13 | 1568 | 149 | |
GenBank mRNA | AB188493.1 | - | 11 - 12 | 1681 | 149 |
AB271694.1 | - | 10 - 11 | 999 | 149 | |
AF387638.1 | - | 10 - 11 | 1017 | 149 | |
AK225766.1 | - | 11 - 12 | 1512 | 149 | |
AK290339.1 | - | 11 - 12 | 1462 | 149 | |
BC008771.2 | - | 11 - 12 | 1069 | 149 | |
BC084540.1 | - | 11 - 12 | 1136 | 149 | |
BT007342.1 | - | 10 - 11 | 999 | 149 | |
X97630.1 | - | 11 - 12 | 1406 | 149 |