The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039469.2 | NP_001034558.2 | 13 - 14 | 1996 | 84 |
NM_001163296.1 | NP_001156768.1 | 13 - 14 | 1996 | 84 | |
NM_001163297.1 | NP_001156769.1 | 13 - 14 | 1993 | 84 | |
NM_004954.4 | NP_004945.4 | 13 - 14 | 1993 | 84 | |
NM_017490.3 | NP_059672.2 | 13 - 14 | 1810 | 84 | |
XM_011544789.1 | XP_011543091.1 | 14 - 15 | 1887 | 84 | |
XM_011544790.1 | XP_011543092.1 | 14 - 15 | 1887 | 84 | |
XM_011544791.1 | XP_011543093.1 | 14 - 15 | 1884 | 84 | |
XM_011544792.1 | XP_011543094.1 | 14 - 15 | 1887 | 84 | |
XM_011544793.1 | XP_011543095.1 | 14 - 15 | 1887 | 84 | |
XM_011544794.1 | XP_011543096.1 | 14 - 15 | 1884 | 84 | |
XM_011544795.1 | XP_011543097.1 | 14 - 15 | 1887 | 84 | |
XM_011544796.1 | XP_011543098.1 | 14 - 15 | 1884 | 84 | |
XM_011544797.1 | XP_011543099.1 | 14 - 15 | 1887 | 84 | |
XM_011544798.1 | XP_011543100.1 | 14 - 15 | 1884 | 84 | |
XM_017017310.1 | XP_016872799.1 | 13 - 14 | 1966 | 84 | |
XM_017017311.1 | XP_016872800.1 | 14 - 15 | 1884 | 84 | |
GenBank mRNA | AB188493.1 | - | 13 - 14 | 2000 | 84 |
AB271694.1 | - | 12 - 13 | 1315 | 84 | |
AF387638.1 | - | 12 - 13 | 1333 | 84 | |
AK225766.1 | - | 13 - 14 | 1828 | 84 | |
AK290339.1 | - | 13 - 14 | 1778 | 84 | |
BC008771.2 | - | 13 - 14 | 1388 | 84 | |
BC084540.1 | - | 13 - 14 | 1452 | 84 | |
BT007342.1 | - | 12 - 13 | 1318 | 84 | |
X97630.1 | - | 13 - 14 | 1722 | 84 |