The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136528.1 | NP_001130000.1 | 1 - 2 | 219 | 92 |
NM_006216.3 | NP_006207.1 | 1 - 2 | 219 | 92 | |
GenBank mRNA | AK225823.1 | - | 1 - 2 | 188 | 92 |
AK312499.1 | - | 1 - 2 | 233 | 92 | |
BC015663.2 | - | 1 - 2 | 176 | 92 |