The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136528.1 | NP_001130000.1 | 7 - 8 | 1319 | 67 |
NM_001136530.1 | NP_001130002.1 | 7 - 8 | 1249 | 67 | |
NM_006216.3 | NP_006207.1 | 7 - 8 | 1322 | 67 | |
NR_073116.1 | 8 - 9 | 1737 | 67 | ||
XM_005246641.2 | XP_005246698.1 | 7 - 8 | 1411 | 67 | |
XM_017004329.1 | XP_016859818.1 | 7 - 8 | 1303 | 67 | |
XM_017004330.1 | XP_016859819.1 | 7 - 8 | 1140 | 67 | |
XM_017004331.1 | XP_016859820.1 | 7 - 8 | 1300 | 67 | |
XM_017004332.1 | XP_016859821.1 | 7 - 8 | 1137 | 67 | |
GenBank mRNA | A03911.1 | - | 6 - 7 | 1100 | 67 |
AK225823.1 | - | 7 - 8 | 1287 | 67 | |
AK295564.1 | - | 7 - 8 | 1217 | 67 | |
AK297588.1 | - | 7 - 8 | 1124 | 67 | |
AK312499.1 | - | 7 - 8 | 1336 | 67 | |
BC015663.2 | - | 7 - 8 | 1279 | 67 | |
BC042628.1 | - | 8 - 9 | 1736 | 67 | |
M17783.1 | - | 6 - 7 | 1076 | 67 |